RIE156328418 Rieke Metals, Inc.

2'-chloro-3-(3-chloro-5-fluorophenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843955
  • Cas Number : 898751-12-7
  • Molecular Weight : 315.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-50
Availability details for
Quantity
  • Availability
  • Add to Cart