RIE156328420 Rieke Metals, Inc.

4'-chloro-3-(3-chloro-5-fluorophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843957
  • Cas Number : 898751-20-7
  • Molecular Weight : 315.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-52
Availability details for
Quantity
  • Availability
  • Add to Cart