RIE156328424 Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-3',4'-dichloropropiophenone

  • MDL number MFCD03843961
  • Cas Number : 898751-36-5
  • Molecular Weight : 331.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5100-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5100-56
Availability details for
Quantity
  • Availability
  • Add to Cart