RIE156328431 Rieke Metals, Inc.

2-(3-chloro-5-fluorophenyl)ethyl cyclopropyl ketone

  • MDL number MFCD03843967
  • Cas Number : 898751-64-9
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5101-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5101-00
Availability details for
Quantity
  • Availability
  • Add to Cart