RIE156328432 Rieke Metals, Inc.

2-(3-Chloro-5-fluorophenyl)ethyl cyclobutyl ketone

  • MDL number MFCD03843968
  • Cas Number : 898751-67-2
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5102-00
Availability details for
Quantity
  • Availability
  • Add to Cart