RIE156328434 Rieke Metals, Inc.

2-(3-chloro-5-fluorophenyl)ethyl cyclohexyl ketone

  • MDL number MFCD03843970
  • Cas Number : 898751-73-0
  • Molecular Weight : 268.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5104-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5104-00
Availability details for
Quantity
  • Availability
  • Add to Cart