RIE156328436 Rieke Metals, Inc.

3-(3,5-Dichlorophenyl)-2',6'-dichloropropiophenone

  • MDL number MFCD08064164
  • Cas Number : 898777-47-4
  • Molecular Weight : 348.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-5700-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-5700-62
Availability details for
Quantity
  • Availability
  • Add to Cart