RIE156328440 Rieke Metals, Inc.

2'-methyl-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844024
  • Cas Number : 898777-58-7
  • Molecular Weight : 278.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-02
Availability details for
Quantity
  • Availability
  • Add to Cart