RIE156328444 Rieke Metals, Inc.

3'-methoxy-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844028
  • Cas Number : 898777-66-7
  • Molecular Weight : 294.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-06
Availability details for
Quantity
  • Availability
  • Add to Cart