RIE156328448 Rieke Metals, Inc.

4'-cyano-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844032
  • Cas Number : 898777-74-7
  • Molecular Weight : 289.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-10
Availability details for
Quantity
  • Availability
  • Add to Cart