RIE156328456 Rieke Metals, Inc.

3'-chloro-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844039
  • Cas Number : 898777-90-7
  • Molecular Weight : 298.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-20
Availability details for
Quantity
  • Availability
  • Add to Cart