RIE156328465 Rieke Metals, Inc.

3',5'-dimethyl-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844048
  • Cas Number : 898778-12-6
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-31
Availability details for
Quantity
  • Availability
  • Add to Cart