RIE156328467 Rieke Metals, Inc.

4'-chloro-3'-fluoro-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844050
  • Cas Number : 898778-18-2
  • Molecular Weight : 316.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-33
Availability details for
Quantity
  • Availability
  • Add to Cart