RIE156328472 Rieke Metals, Inc.

3'-trifluoromethyl-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844055
  • Cas Number : 898778-33-1
  • Molecular Weight : 332.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-44
Availability details for
Quantity
  • Availability
  • Add to Cart