RIE156328473 Rieke Metals, Inc.

4'-trifluoromethyl-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844056
  • Cas Number : 898778-36-4
  • Molecular Weight : 332.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-45
Availability details for
Quantity
  • Availability
  • Add to Cart