RIE156328482 Rieke Metals, Inc.

3',5'-dichloro-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844065
  • Cas Number : 898778-60-4
  • Molecular Weight : 333.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-57
Availability details for
Quantity
  • Availability
  • Add to Cart