RIE156328486 Rieke Metals, Inc.

3',4',5'-trifluoro-3-(3,4,5-trifluorophenyl)propiophenone

  • MDL number MFCD03844069
  • Cas Number : 898778-68-2
  • Molecular Weight : 318.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6100-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6100-61
Availability details for
Quantity
  • Availability
  • Add to Cart