RIE156328491 Rieke Metals, Inc.

cyclohexyl 2-(3,4,5-trifluorophenyl)ethyl ketone

  • MDL number MFCD03844073
  • Cas Number : 898778-77-3
  • Molecular Weight : 270.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-6104-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-6104-00
Availability details for
Quantity
  • Availability
  • Add to Cart