RIE156328492 Rieke Metals, Inc.

3'-(1,3-Dioxolan-2-yl)-2-methylbenzophenone

  • MDL number MFCD07699946
  • Cas Number : 898778-81-9
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-02
Availability details for
Quantity
  • Availability
  • Add to Cart