RIE156328493 Rieke Metals, Inc.

3-(1,3-Dioxolan-2-yl)-3'-methylbenzophenone

  • MDL number MFCD07699947
  • Cas Number : 898778-83-1
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-03
Availability details for
Quantity
  • Availability
  • Add to Cart