RIE156328495 Rieke Metals, Inc.

3'-(1,3-Dioxolan-2-yl)-2-methoxybenzophenone

  • MDL number MFCD07699949
  • Cas Number : 898778-87-5
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-05
Availability details for
Quantity
  • Availability
  • Add to Cart