RIE156328497 Rieke Metals, Inc.

3-(1,3-Dioxolan-2-yl)-4'-methoxybenzophenone

  • MDL number MFCD07699951
  • Cas Number : 898778-91-1
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-07
Availability details for
Quantity
  • Availability
  • Add to Cart