RIE156328498 Rieke Metals, Inc.

2-cyano-3'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699952
  • Cas Number : 898778-93-3
  • Molecular Weight : 279.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-08
Availability details for
Quantity
  • Availability
  • Add to Cart