RIE156328503 Rieke Metals, Inc.

4'-carboethoxy-3-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699957
  • Cas Number : 898779-06-1
  • Molecular Weight : 326.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-13
Availability details for
Quantity
  • Availability
  • Add to Cart