RIE156328505 Rieke Metals, Inc.

3-(1,3-dioxolan-2-yl)-4'-thiomethylbenzophenone

  • MDL number MFCD07699959
  • Cas Number : 898779-12-9
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-15
Availability details for
Quantity
  • Availability
  • Add to Cart