RIE156328509 Rieke Metals, Inc.

4'-Chloro-3-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699963
  • Cas Number : 741707-93-7
  • Molecular Weight : 288.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-21
Availability details for
Quantity
  • Availability
  • Add to Cart