RIE156328515 Rieke Metals, Inc.

2,6-dimethyl-3'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699969
  • Cas Number : 898779-39-0
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-29
Availability details for
Quantity
  • Availability
  • Add to Cart