RIE156328516 Rieke Metals, Inc.

3,4-dimethyl-3'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699970
  • Cas Number : 898779-42-5
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-30
Availability details for
Quantity
  • Availability
  • Add to Cart