RIE156328520 Rieke Metals, Inc.

3-chloro-3'-(1,3-dioxolan-2-yl)-4-fluorobenzophenone

  • MDL number MFCD07699974
  • Cas Number : 898759-22-3
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-34
Availability details for
Quantity
  • Availability
  • Add to Cart