RIE156328523 Rieke Metals, Inc.

3'-(1,3-dioxolan-2-yl)-2-trifluoromethylbenzophenone

  • MDL number MFCD07699977
  • Cas Number : 898759-31-4
  • Molecular Weight : 322.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-43
Availability details for
Quantity
  • Availability
  • Add to Cart