RIE156328529 Rieke Metals, Inc.

4-chloro-3'-(1,3-dioxolan-2-yl)-2-fluorobenzophenone

  • MDL number MFCD07699983
  • Cas Number : 898759-46-1
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-52
Availability details for
Quantity
  • Availability
  • Add to Cart