RIE156328533 Rieke Metals, Inc.

3,4-Dichloro-3'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699987
  • Cas Number : 898759-58-5
  • Molecular Weight : 323.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-56
Availability details for
Quantity
  • Availability
  • Add to Cart