RIE156328535 Rieke Metals, Inc.

2,4-difluoro-3'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07699989
  • Cas Number : 898759-63-2
  • Molecular Weight : 290.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-58
Availability details for
Quantity
  • Availability
  • Add to Cart