RIE156328538 Rieke Metals, Inc.

3-(1,3-dioxolan-2-yl)-3',4',5'-trifluorobenzophenone

  • MDL number MFCD07699992
  • Cas Number : 898759-70-1
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-61
Availability details for
Quantity
  • Availability
  • Add to Cart