RIE156328539 Rieke Metals, Inc.

Cyclopropyl 3-(1,3-dioxolan-2-yl)phenyl ketone

  • MDL number MFCD07699993
  • Cas Number : 898759-72-3
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-01-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-01-00
Availability details for
Quantity
  • Availability
  • Add to Cart