RIE156328542 Rieke Metals, Inc.

cyclohexyl 3-(1,3-dioxolan-2-yl)phenyl ketone

  • MDL number MFCD07699996
  • Cas Number : 898759-78-9
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-04-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-04-00
Availability details for
Quantity
  • Availability
  • Add to Cart