RIE156328546 Rieke Metals, Inc.

4-(1,3-dioxolan-2-yl)-4'-methylbenzophenone

  • MDL number MFCD07699999
  • Cas Number : 898759-84-7
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-04
Availability details for
Quantity
  • Availability
  • Add to Cart