RIE156328548 Rieke Metals, Inc.

4'-(1,3-dioxolan-2-yl)-3-methoxybenzophenone

  • MDL number MFCD07700001
  • Cas Number : 898759-88-1
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-06
Availability details for
Quantity
  • Availability
  • Add to Cart