RIE156328549 Rieke Metals, Inc.

4-(1,3-dioxolan-2-yl)-4'-methoxybenzophenone

  • MDL number MFCD07700002
  • Cas Number : 898759-90-5
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-07
Availability details for
Quantity
  • Availability
  • Add to Cart