RIE156328550 Rieke Metals, Inc.

2-cyano-4'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07700003
  • Cas Number : 898759-92-7
  • Molecular Weight : 279.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-08
Availability details for
Quantity
  • Availability
  • Add to Cart