RIE156328563 Rieke Metals, Inc.

4-(1,3-Dioxolan-2-yl)-4'-fluorobenzophenone

  • MDL number MFCD07700016
  • Cas Number : 898760-14-0
  • Molecular Weight : 272.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-23
Availability details for
Quantity
  • Availability
  • Add to Cart