RIE156328566 Rieke Metals, Inc.

2,5-dimethyl-4'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07700019
  • Cas Number : 898760-22-0
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-28
Availability details for
Quantity
  • Availability
  • Add to Cart