RIE156328567 Rieke Metals, Inc.

2,6-dimethyl-4'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07700020
  • Cas Number : 898760-25-3
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-29
Availability details for
Quantity
  • Availability
  • Add to Cart