RIE156328568 Rieke Metals, Inc.

3,4-dimethyl-4'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07700021
  • Cas Number : 898760-28-6
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-30
Availability details for
Quantity
  • Availability
  • Add to Cart