RIE156328570 Rieke Metals, Inc.

4-bromo-4'-(1,3-dioxolan-2-yl)-3-fluorobenzophenone

  • MDL number MFCD07700023
  • Cas Number : 898760-34-4
  • Molecular Weight : 351.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-32
Availability details for
Quantity
  • Availability
  • Add to Cart