RIE156328574 Rieke Metals, Inc.

4'-(1,3-dioxolan-2-yl)-2-fluorobenzophenone

  • MDL number MFCD07700027
  • Cas Number : 898760-46-8
  • Molecular Weight : 272.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-37
Availability details for
Quantity
  • Availability
  • Add to Cart