RIE156328576 Rieke Metals, Inc.

4'-(1,3-dioxolan-2-yl)-3-trifluoromethylbenzophenone

  • MDL number MFCD07700029
  • Cas Number : 898760-52-6
  • Molecular Weight : 322.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-44
Availability details for
Quantity
  • Availability
  • Add to Cart