RIE156328589 Rieke Metals, Inc.

3,5-difluoro-4'-(1,3-dioxolan-2-yl)benzophenone

  • MDL number MFCD07700042
  • Cas Number : 898760-80-0
  • Molecular Weight : 290.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-00-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-00-60
Availability details for
Quantity
  • Availability
  • Add to Cart