RIE156328592 Rieke Metals, Inc.

cyclobutyl 4-(1,3-dioxolan-2-yl)phenyl ketone

  • MDL number MFCD07700045
  • Cas Number : 898760-86-6
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-02-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-02-00
Availability details for
Quantity
  • Availability
  • Add to Cart