RIE156328595 Rieke Metals, Inc.

1-(4-bromo-3-methylphenyl)-5-chloro-1-oxopentane

  • MDL number MFCD03844177
  • Cas Number : 898760-94-6
  • Molecular Weight : 289.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6130D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6130D
Availability details for
Quantity
  • Availability
  • Add to Cart